Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1,2-Dichloroethene Solution, 5000μg/mL in Methanol, Ultra Scientific

Catalog No. USEPA1028
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-EPA-1028 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-EPA-1028 Supplier Agilent Technologies Supplier No. EPA10281
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 156-59-2 , 67-56-1
Molecular Formula C2H2Cl2
Molecular Weight (g/mol) 96.938
InChI Key KFUSEUYYWQURPO-UPHRSURJSA-N
PubChem CID 643833
ChEBI CHEBI:28805
IUPAC Name (Z)-1,2-dichloroethene
SMILES C(=CCl)Cl

Specifications

Physical Form Liquid
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 5000μg/mL
Chemical Name or Material cis-1, 2-Dichloroethene, 5000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.