Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1,3-Cyclohexanedicarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. C219725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C219725G 25 g
C21975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C219725G Supplier TCI America Supplier No. C219725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2305-31-9
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00134411
InChI Key XBZSBBLNHFMTEB-OLQVQODUSA-N
PubChem CID 6432255
IUPAC Name (1S,3R)-cyclohexane-1,3-dicarboxylic acid
SMILES C1CC(CC(C1)C(=O)O)C(=O)O

Specifications

Melting Point 165°C
Color White
Quantity 25 g
Formula Weight 172.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material cis-1,3-Cyclohexanedicarboxylic Acid
EINECSNumber (3)-2685
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.