Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1,4-Cyclohexanediamine 98.0+%, TCI America™
SDP

Catalog No. C17985G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17985G 5 g
C179825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. C17985G Supplier TCI America Supplier No. C17985G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 15827-56-2
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00035862,MFCD00075174,MFCD08276323
InChI Key VKIRRGRTJUUZHS-UHFFFAOYSA-N
Synonym cis-1,4-Diaminocyclohexane
PubChem CID 18374
IUPAC Name cyclohexane-1,4-diamine
SMILES NC1CCC(N)CC1

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
UN Number 2735
Formula Weight 114.19
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material cis-1,4-Cyclohexanediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.