Learn More
AdipoGen cis-1 4-Dichloro-2-butene
SDP
Supplier:  AdipoGen CDXC0779G025
Encompass_Preferred
Chemical. CAS 1476-11-5. Formula C4H6Cl2. MW 125. cis-1,4-Dichloro-2-butene is a bulding block and intermediate used in the synthesis and for the production of other chemicals such as acrylonitrile, adiponitrile, butane-1,4-diol, chloroprene and tetrahydrofuran. It undergoes ring opening to give vinyl chloride and 1,4 dichlorobutane. The cis configuration has lower activation energies than the trans configuration so it is more stable thermodynamically.
Catalog No. 50-256-1338
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.