Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-cis-2-Benzylaminocyclohexanemethanol 98.0+%, TCI America™

Catalog No. B11195G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11195G 5 g
B11191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11195G Supplier TCI America Supplier No. B11195G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 71581-92-5
Molecular Formula C14H21NO
Molecular Weight (g/mol) 219.328
MDL Number MFCD00151395
InChI Key BRQFIORUNWWNBM-KBPBESRZSA-N
Synonym 1r,2s-2-benzylamino cyclohexyl methanol, +-cis-2-benzylaminocyclohexanemethanol, cis-1r,2s-+-2-benzylamino cyclohexanemethanol, --cis-2-benzylaminocyclohexanemethanol, cis-1r,2s-+-2-benzylaminocyclohexanemethanol, pubchem7939, cis-2-benzylamino cyclohexylmethanol, 2-benzylamino cyclohexyl methanol #, cis-2-benzylamino cyclohexyl methanol
PubChem CID 853687
IUPAC Name [(1R,2S)-2-(benzylamino)cyclohexyl]methanol
SMILES C1CCC(C(C1)CO)NCC2=CC=CC=C2

Specifications

Melting Point 68°C
Quantity 5 g
Formula Weight 219.33
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (+)-cis-2-Benzylaminocyclohexanemethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.