Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid, 97%

Catalog No. AAH6232903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6232903 1 g
AAH62329MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6232903 Supplier Thermo Scientific Chemicals Supplier No. H6232903
Add to Cart

CAS: 685508-28-5 | C13H15NO4 | 249.266 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 685508-28-5
Molecular Formula C13H15NO4
Molecular Weight (g/mol) 249.266
MDL Number MFCD03844615
InChI Key ZNRUEEQIUVKKBL-MNOVXSKESA-N
Synonym cis-2-benzyloxycarbonylaminocyclobutane-carboxylic acid, cis-2-bocamino cyclobutanecarboxylic acid, cis-2-cbz-amino cyclobutane-1-carboxylic acid, 2beta-benzyloxycarbonylamino cyclobutane-1beta-carboxylic acid, 1r,2s-2-benzyloxy carbonyl amino cyclobutane-1-carboxylic acid
PubChem CID 68498468
IUPAC Name (1R,2S)-2-(phenylmethoxycarbonylamino)cyclobutane-1-carboxylic acid
SMILES C1CC(C1C(=O)O)NC(=O)OCC2=CC=CC=C2

Specifications

Quantity 1 g
Formula Weight 249.26
Percent Purity 97%
Chemical Name or Material cis-2-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.