Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-2-Hexen-1-ol, 94%, remainder mainly trans isomer

Catalog No. AAA1373406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1373406 5 g
AAA1373414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1373406 Supplier Thermo Scientific Chemicals Supplier No. A1373406
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 928-94-9 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 928-94-9
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00063209
InChI Key ZCHHRLHTBGRGOT-PLNGDYQASA-N
Synonym cis-2-hexen-1-ol, 2-hexen-1-ol, z, cis-hex-2-en-1-ol, z-2-hexen-1-ol, z-hex-2-en-1-ol, unii-871tel510e, 2z-hexen-1-ol, 2z-hex-2-en-1-ol, cis-1-hydroxy-2-hexene, cis-2-hexenol
PubChem CID 5324489
IUPAC Name (Z)-hex-2-en-1-ol
SMILES CCC\C=C/CO

Specifications

Density 0.847
Boiling Point 158°C
Flash Point 54°C (129°F)
Refractive Index 1.441
Quantity 5 g
UN Number UN1987
Beilstein 1719708
Formula Weight 100.16
Percent Purity 94%
Assay remainder mainly trans isomer
Chemical Name or Material cis-2-Hexen-1-ol
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226-H319
Flammable liquid and vapour.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 213-190-7
RTECSNumber MP8395000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.