Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-2-Octene 95.0+%, TCI America™
SDP

Catalog No. O00981ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O00981ML 1 mL
O00985ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O00981ML Supplier TCI America Supplier No. O00981ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7642-04-8
Molecular Formula C8H16
Molecular Weight (g/mol) 112.22
MDL Number MFCD00065146
InChI Key ILPBINAXDRFYPL-HWKANZROSA-N
PubChem CID 5364448
IUPAC Name (E)-oct-2-ene
SMILES CCCCCC=CC

Specifications

Color Colorless
Boiling Point 126°C
Quantity 1 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-2-Octene
EINECSNumber (2)-0024
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.