Learn More
Sigma Aldrich Fine Chemicals Biosciences cis 3 Hexen 1 ol natural 9
Supplier:  Sigma Aldrich Fine Chemicals Biosciences W2563234KGK
Encompass
cis-3-Hexen-1-ol is one of the key volatile constituents of green leaf volatiles(GLV) that can act as an attractant to various insects. It is emitted by green plants when they are physically damaged.
Catalog No. 50-291-4359
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.