Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-4-(Benzyloxycarbonylamino)cyclohexanecarboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog No. AAH6254603
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6254603 1 g
AAH62546MD 250 mg
AAH6254606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6254603 Supplier Thermo Scientific Chemicals Supplier No. H6254603
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 917022-26-5
Molecular Formula C15H19NO3
Molecular Weight (g/mol) 261.321
MDL Number MFCD03844593
InChI Key SOIRQISQWQDGSN-UHFFFAOYSA-N
PubChem CID 21882542
IUPAC Name benzyl N-(4-formylcyclohexyl)carbamate
SMILES C1CC(CCC1C=O)NC(=O)OCC2=CC=CC=C2

Specifications

Synonym benzyl 4-formylcyclohexylcarbamate, benzyl cis-4-formylcyclohexyl carbamate, benzyl cis-4-formylcyclohexylcarbamate, benzyl 4-formylcyclohexyl carbamate, trans-4-benzyloxycarbonylamino-cyclohexanecarbaldehyde, benzyl 1r,4r-4-formylcyclohexylcarbamate, trans-4-benzyloxycarbonylamino cyclohexanecarbaldehyde, benzyl n-4-formylcyclohexyl carbamate, benzyl4-formylcyclohexylcarbamate, soirqisqwqdgsn-mqmhxkeqsa-n
Quantity 1 g
Formula Weight 261.31
Percent Purity 97%
Chemical Name or Material cis-4-(Benzyloxycarbonylamino)cyclohexanecarboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.