Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-4-(Boc-amino)cyclohexanemethylamine, 97%

Catalog No. AAH6218703
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6218703 1 g
AAH62187MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6218703 Supplier Thermo Scientific Chemicals Supplier No. H6218703
Add to Cart

CAS: 509143-00-4 | C12H24N2O2 | 228.336 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 509143-00-4
Molecular Formula C12H24N2O2
Molecular Weight (g/mol) 228.336
MDL Number MFCD03844602
InChI Key NVQFOBONHIXDOC-UHFFFAOYSA-N
Synonym tert-butyl trans-4-aminomethyl cyclohexyl carbamate, tert-butyl n-4-aminomethyl cyclohexyl carbamate, tert-butyl cis-4-aminomethylcyclohexylcarbamate, tert-butyl n-1r,4r-4-aminomethyl cyclohexyl carbamate, tert-butyl trans-4-aminomethylcyclohexyl carbamate, tert-butyl cis-4-aminomethyl cyclohexyl carbamate, cis-4-boc-amino cyclohexylmethylamine, tert-butyl trans-4-aminomethylcyclohexylcarbamate, trans-4-boc-amino cyclohexylmethylamine, tert-butyl 4-aminomethyl cyclohexyl carbamate
PubChem CID 2756045
IUPAC Name tert-butyl N-[4-(aminomethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)CN

Specifications

Quantity 1 g
Formula Weight 228.33
Percent Purity 97%
Chemical Name or Material cis-4-(Boc-amino)cyclohexanemethylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.