Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-4-Hepten-1-ol 95.0+%, TCI America™
SDP

Catalog No. H083325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H083325ML 25 mL
H08335ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H083325ML Supplier TCI America Supplier No. H083325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6191-71-5
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00015575
InChI Key CUKAXHVLXKIPKF-ARJAWSKDSA-N
Synonym cis-4-hepten-1-ol, z-4-hepten-1-ol, z-hept-4-en-1-ol, cis-hept-4-enol, unii-802k6ztp9q, 4z-hept-4-en-1-ol, 4-hepten-1-ol, cis, 4z-4-hepten-1-ol, 4-hepten-1-ol, 4z, 4-hepten-1-ol, z
PubChem CID 5367536
IUPAC Name (Z)-hept-4-en-1-ol
SMILES CCC=CCCCO

Specifications

Color Colorless
Boiling Point 85°C
Quantity 25 mL
Formula Weight 114.19
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-4-Hepten-1-ol
EINECSNumber (2)-2393
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.