Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-4-Nitrostilbene 97.0+%, TCI America™
SDP

Catalog No. N10851G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10851G 1 g
N1085200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N10851G Supplier TCI America Supplier No. N10851G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6624-53-9
Molecular Formula C14H11NO2
Molecular Weight (g/mol) 225.247
InChI Key ZISCOWXWCHUSMH-SREVYHEPSA-N
Synonym (Z)-4-Nitrostilbene
PubChem CID 1551577
IUPAC Name 1-nitro-4-[(Z)-2-phenylethenyl]benzene
SMILES C1=CC=C(C=C1)C=CC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 63°C
Quantity 1 g
Formula Weight 225.25
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material cis-4-Nitrostilbene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.