Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-4-tert-Butylcyclohexanecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. B263125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B263125G 25 g
B26315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B263125G Supplier TCI America Supplier No. B263125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 943-28-2
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
InChI Key QVQKEGYITJBHRQ-UHFFFAOYSA-N
PubChem CID 136759
IUPAC Name 4-tert-butylcyclohexane-1-carboxylic acid
SMILES CC(C)(C)C1CCC(CC1)C(=O)O

Specifications

Melting Point 117°C
Color White
Quantity 25 g
Formula Weight 184.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material cis-4-tert-Butylcyclohexanecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.