Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-Diammineplatinum(II) Dichloride, TCI America™
SDP

Catalog No. D33711G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33711G 1 g
D3371100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33711G Supplier TCI America Supplier No. D33711G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15663-27-1
Molecular Formula Cl2H6N2Pt
Molecular Weight (g/mol) 300.04
MDL Number MFCD00011623
InChI Key BSJGASKRWFKGMV-UHFFFAOYSA-L
Synonym Cisplatin, cis-Diamminedichloroplatinum(II), cis-Platinum(II) Diammine Dichloride
IUPAC Name dichloroplatinumbis(ylium) diamine
SMILES N.N.Cl[Pt++]Cl

Specifications

Melting Point 270°C
Color Yellow
Quantity 1 g
UN Number 3288
Formula Weight 300.05
Physical Form Crystalline Powder
Chemical Name or Material cis-Diammineplatinum(II) Dichloride
RTECSNumber TP2450000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.