accessibility menu, dialog, popup

UserName

Viewing profile as user:
NClNClPt

CAS RN 15663-27-1

IUPAC Name: dichloroplatinumbis(ylium) diamine
Synonyms: plastin|Cis-DDP|platinol-AQ|CACP|cis pt II|CPDC|Trans-DDP|CPDD|cisplatyl|platidiam
Molecular Weight (g/mol): 300.04
Molecular Formula: Cl2H6N2Pt
InChi Key: BSJGASKRWFKGMV-UHFFFAOYSA-L
SMILES: N.N.Cl[Pt++]Cl
Molecular Weight (g/mol) 
  • (8)
  • (3)
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (3)
  • (3)
Grade 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (6)
  • (1)
  • (1)
  • (8)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)