Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-Jasmone, 94%, Thermo Scientific™

Catalog No. AAA1906014
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1906014 25 g
AAA1906006 5 g
AAA1906022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1906014 Supplier Thermo Scientific Chemicals Supplier No. A1906014

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 488-10-8
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.25
MDL Number MFCD00001402,MFCD00001402,MFCD00001402
InChI Key XMLSXPIVAXONDL-PLNGDYQASA-N
Synonym trans-jasmone, e-jasmone, unii-7tcs3y45dr, 7tcs3y45dr, 3-methyl-2-e-pent-2-enyl cyclopent-2-en-1-one, 2-cyclopenten-1-one, 3-methyl-2-2-pentenyl-, e, 3-methyl-2-pent-2-en-1-yl cyclopent-2-en-1-one, 3-methyl-2-2e-pent-2-en-1-yl cyclopent-2-en-1-one, jasmone, e, 3-methyl-2-2-pentenyl-2-cyclopenten-1-one
PubChem CID 1549019
IUPAC Name 3-methyl-2-[(2Z)-pent-2-en-1-yl]cyclopent-2-en-1-one
SMILES CC\C=C/CC1=C(C)CCC1=O

Specifications

Density 0.94
Boiling Point 134°C to 135°C (12 mmHg)
Flash Point 107°C (224°F)
Odor Pleasant
Refractive Index 1.499
Quantity 25 g
Beilstein 1907713
Merck Index 14,5259
Formula Weight 164.25
Percent Purity 94%
Chemical Name or Material cis-Jasmone
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 207-668-4
RTECSNumber GY7301000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.