Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-Ned 19, Tocris Bioscience™

Catalog No. 395310
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-531-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 39-531-0 Supplier R&D Systems Supplier No. 3953/10
This item is not returnable. View return policy

NAADP antagonist; inhibits Ca2+ release

Irreversible nicotinic acid adenine dinucleotide phosphate (NAADP) antagonist; inhibits Ca2+ release (IC50 = 800 nM) and [32P]NAADP binding (IC50 = 15 μM). Stereoisomer of trans-Ned 19 . Fluorescently labels NAADP receptors in intact cells (excitation 351 and 365 nm using an ultraviolet Argon laser).

Isomer of trans-Ned 19 .

Chemical Identifiers

CAS 1137264-00-6
Molecular Formula C30H31FN4O3
Molecular Weight (g/mol) 514.601
InChI Key FUHCEERDBRGPQZ-LSYYVWMOSA-N
Synonym cis-ned 19, 1s,3s-1-3-4-2-fluorophenyl piperazin-1-yl methyl-4-methoxyphenyl-2,3,4,9-tetrahydro-1h-pyrido 3,4-b indole-3-carboxylic acid
PubChem CID 1427626
IUPAC Name (1S,3S)-1-[3-[[4-(2-fluorophenyl)piperazin-1-yl]methyl]-4-methoxyphenyl]-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic acid
SMILES COC1=C(C=C(C=C1)C2C3=C(CC(N2)C(=O)O)C4=CC=CC=C4N3)CN5CCN(CC5)C6=CC=CC=C6F

Specifications

Quantity 10 mg
Formula Weight 514.59
Percent Purity >98%
Chemical Name or Material cis-Ned 19
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.