Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-Nonachlor Solution, 100μg/mL in methanol, Ultra Scientific

Catalog No. USPP4901
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-PP-4901 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-PP-4901 Supplier Agilent Technologies Supplier No. PP4901
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5103-73-1 , 67-56-1
Molecular Formula C10H5Cl9
Molecular Weight (g/mol) 444.2
InChI Key OCHOKXCPKDPNQU-MSFPVELZSA-N
PubChem CID 12313426
SMILES C12C(C(C(C1Cl)Cl)Cl)C3(C(=C(C2(C3(Cl)Cl)Cl)Cl)Cl)Cl

Specifications

Physical Form Liquid
Color Colorless
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material cis-Nonachlor Solution, 100μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.