Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-Nonachlor, SPEX CertiPrep™
SDP

Catalog No. S2765 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S-2765 1 mL 2mL amber prescored glass ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S-2765 Supplier SPEX CertiPrep Supplier No. S2765

missing translation for 'validMainframeMsg'

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
cis-Nonachlor5103-73-11000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 5103-73-1 , 67-56-1
Molecular Formula C10H5Cl9
Molecular Weight (g/mol) 444.2
InChI Key OCHOKXCPKDPNQU-MSFPVELZSA-N
Synonym cis-nonachlor, cis-nonachlor solution, cis-nonachlor standard, trans-nonachlor solution, trans-nonachlor standard, cis-nonachlor 10 microg/ml in cyclohexane, cis-nonachlor 100 microg/ml in isooctane, trans-nonachlor 10 microg/ml in cyclohexane, trans-nonachlor 100 microg/ml in cyclohexane, cis-nonachlor solution, 100 mug/ml in hexane, analytical standard
PubChem CID 12313426
SMILES C12C(C(C(C1Cl)Cl)Cl)C3(C(=C(C2(C3(Cl)Cl)Cl)Cl)Cl)Cl

Specifications

CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
Packaging 2mL amber prescored glass ampule
Quantity 1 mL
Chemical Name or Material cis-Nonachlor
Hazard Category Highly flammable liquid and vapor. Toxic if inhaled. Causes damage to organs.
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.