Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-Octahydroisoindole 97.0+%, TCI America™
SDP

Catalog No. O04711G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04711G 1 g
O04715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04711G Supplier TCI America Supplier No. O04711G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1470-99-1
Molecular Formula C8H15N
Molecular Weight (g/mol) 125.215
MDL Number MFCD07778620
InChI Key ODSNARDHJFFSRH-OCAPTIKFSA-N
Synonym cis-octahydroisoindole, cis-octahydro-1h-isoindole, cis-hexahydroisoindoline, 3ar,7as-octahydro-1h-isoindole, 1h-isoindole,octahydro-, 3ar,7as, 1h-isoindole, octahydro-, 3ar,7as-rel, 6s,1r-8-azabicyclo 4.3.0 nonane, 3as,7ar-octahydro-1h-isoindole, 3as,7ar-octahydro-isoindole, cis-octahyroisoindole
PubChem CID 10964494
IUPAC Name (3aS,7aR)-2,3,3a,4,5,6,7,7a-octahydro-1H-isoindole
SMILES C1CCC2CNCC2C1

Specifications

Color Yellow
Boiling Point 189°C
Quantity 1 g
Formula Weight 125.22
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material cis-Octahydroisoindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.