Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Cis-Resveratrol, Antioxidant Agent, >98% Purity, 50 mg

Catalog No. 502705728
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5728 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5728 Supplier Abcam Supplier No. AB14443650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antioxidant agent and NADPH oxidase inhibitor that blocks ROS production.

  • Purity: >98%
  • Molecular Weight: 228.24 Da
  • Biological Activity: Antioxidant agent and NADPH oxidase inhibitor that blocks ROS production
  • Key Effects: Inhibits canonical and non-canonical inflammasomes in macrophages; shows anti-inflammatory and antiproliferative effects
  • Additional Information: Resveratrol (ab120726) isomer; orally bioavailable

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule NADPH Oxidase
Product Type Antioxidant
Buffer 95% ethanol, 5% 5-[(Z)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol
Content And Storage Store at -80°C.
Form Liquid
Shipping Condition Dry Ice
Substrate Properties Chemical Substrate
Formulation 95% Ethanol, 5% 5-[(Z)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol
Quantity 50 mg
For Use With (Application) Antioxidant, NADPH oxidase inhibition, Inflammasome inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.