Learn More
Abcam Cis-Resveratrol, Antioxidant Agent, >98% Purity, Liquid, 10 mg
Description
- Molecular Weight: 228.24 Da
- Purity: >98%
- Form: Liquid
- CAS Number: 61434-67-1
- Biological Activity: Antioxidant agent, NADPH oxidase inhibitor; blocks ROS production; isomer of resveratrol (ab120726); inhibits canonical and non-canonical inflammasomes in macrophages; exhibits anti-inflammatory and antiproliferative effects
- Bioavailability: Orally bioavailable
- Storage Buffer: 95% Ethanol, 5% 5-[(Z)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol
- Storage Temperature: Store at -80°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Product Type | Antioxidant |
| Form | Liquid |
| Buffer | 95% ethanol, 5% 5-[(Z)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol |
| Content And Storage | Store at -80°C. |
| Shipping Condition | Dry Ice |
| Substrate Properties | Chemical Substrate |
| Target Molecule | NADPH Oxidase |
| Formulation | 95% Ethanol, 5% 5-[(Z)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol |
| Quantity | 10 mg |
| For Use With (Application) | Antioxidant studies, NADPH oxidase inhibition, Inflammasome research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.