Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Cis-Resveratrol, Antioxidant Agent, Greater Than 98 Percent Purity, 5 mg

Catalog No. 502705727
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5727 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5727 Supplier Abcam Supplier No. AB1444365MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent antioxidant agent and NADPH oxidase inhibitor with anti-inflammatory and antiproliferative effects.

  • Molecular weight: 228.24 Da
  • Purity: >98%
  • Form: Liquid
  • Storage buffer: Constituents: 95% Ethanol, 5% 5-[(Z)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol
  • Biological activity: Antioxidant agent, NADPH oxidase inhibitor that blocks ROS production, and inhibitor of canonical and non-canonical inflammasomes in macrophages
  • Effects: Anti-inflammatory and antiproliferative
  • Pharmacokinetics: Orally bioavailable
  • Note: Isomer of Resveratrol (ab120726)

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule NADPH Oxidase
Shipping Condition Dry Ice
Product Type Antioxidant
Buffer 95% ethanol, 5% 5-[(Z)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol
Content And Storage Store at -80°C.
Form Liquid
Formulation 95% Ethanol, 5% 5-[(Z)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol
Quantity 5 mg
For Use With (Application) Antioxidant studies, NADPH oxidase inhibition, Inflammasome research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.