Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Cisapride monohydrate

Catalog No. AC465900010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465900010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC465900010 Supplier Thermo Scientific Chemicals Supplier No. 465900010
Add to Cart

CAS: 260779-88-2 | C23H29ClFN3O4·H2O, C23H31ClFN3O5 | 483.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 260779-88-2
Molecular Formula C23H29ClFN3O4·H2O

Specifications

Quantity 1 g
Formula Weight 483.97
Chemical Name or Material Cisapride monohydrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes serious eye damage.

Precautionary Statement GHS P Statement Wear eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.