Learn More
Cayman Chemical Cisplatin, 15663-27-1, MFCD00011623, 50mg
SDP
Supplier:  Cayman Chemical 1311950
Encompass_Preferred
Molecular Formula: Cl2H6N2Pt, Purity: ≥95%, Formula Weight: 300.1, Synonyms: CDDP, cis-Diamminedichloroplatinum, Cisplatinum, NSC 119875; Cisplatin is a platinum-containing compound that acts as a DNA-crosslinking agent and interferes with replication and transcription, culminating in apoptosis.
Catalog No. NC1706394
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.