Learn More
Enzo Life Sciences (+)-cis,trans-Abscisic acid (1mg). CAS: 21293-29-8
SDP
Supplier:  Enzo Life Sciences ALX350255M001
Encompass_Preferred
Natural and active isomer of the abscission accelerating plant hormone. Alternative name: (S)-5-(1-Hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl)-3-methyl-[2Z,4E]-pentadienoic acid, (+)-cis,trans ABA. Purity: ≥97% (HPLC). Solubility: Soluble in ethanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8806
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.