Learn More
CPC Scientific H-Ala-Thr-Lys-Ala-Ala-Cit-Lys-Ser-Ala-Pro-Ala-Thr-Gly-Gly-Val-Lys-Lys-Pro-His-Arg-Tyr-Arg-Pro-Gly-Gly-Gly-Lys(Biotin)-OH (trifluoroacetate salt) 0.25MG
SDP
Supplier:  CPC Scientific HIST057A
Encompass
This peptide is the fragment of Histone H3 (21-44) with deimination at Arg26, converting it to Cit (citrulline). It is biotinylated through a C-terminal GGK linker. Deimination by peptidyl arginine deiminase 4 (PADI4) blocks methylation by the CARM1 methyltransferase and inhibits transcriptional activation.ONE-LETTER SEQUENCE: ATKAA-Cit-KSAPATGGVKKPHRYRPGGG-K(Biotin)MOLECULAR FORMULA: MOLECULAR WEIGHT:2975.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Cuthbert, G. et al. Cell 118, 545 (2004)
Catalog No. 50-210-9505
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.