Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Citalopram, 5-HT Reuptake Inhibitor, >98% Purity, Solid, 100 mg

Catalog No. 502584681
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4681 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4681 Supplier Abcam Supplier No. AB120133100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selective serotonin reuptake inhibitor with greater than 98% purity.

  • Molecular weight: 405.3 Da
  • Purity: >98%
  • Description: Selective serotonin reuptake inhibitor. Antidepressant in vivo.
  • Form: Solid
  • CAS number: 59729-32-7
  • Storage: Store at ambient temperature
  • Citation count: 5

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Serotonin Transporter
Shipping Condition Ambient
Content And Storage Store at ambient temperature.
Form Solid
Formulation 100% 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile hydrobromide
Quantity 100 mg
For Use With (Application) Antidepressant, 5-HT reuptake inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.