Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Citalopram, 5-HT Reuptake Inhibitor, >98% Purity, Solid, 25 mg

Catalog No. 502584680
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4680 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4680 Supplier Abcam Supplier No. AB12013325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selective serotonin reuptake inhibitor with greater than 98% purity for antidepressant studies in vivo.

  • Molecular Weight: 405.3 Da
  • Purity: >98%
  • Description: Selective serotonin reuptake inhibitor, acts as an antidepressant in vivo
  • Form: Solid
  • Chemical Name: 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile hydrobromide
  • Storage: Store at ambient
  • Shipping: Ambient - Can ship with ice
  • Citation Count: 5

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule 5-HT (Serotonin)
Substrate Properties Chemical Substrate
Content And Storage Store at ambient temperature.
Shipping Condition Ambient
Form Solid
Formulation 100% 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile hydrobromide
Quantity 25 mg
For Use With (Application) Antidepressant research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.