Learn More
CITED1 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
The CITED1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human CITED1. This antibody recognizes human, rat, mouse antigen. The CITED1 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specifications
Specifications
| Antigen | CITED1 |
| Applications | Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Classification | Polyclonal |
| Conjugate | CoraLite 488 |
| Host Species | Rabbit |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 4435 |
| Target Species | Human, Rat, Mouse |
| Isotype | IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.