Learn More
Invitrogen™ CITED2 Monoclonal Antibody (JA22)

Description
Suggested positive control: Hela whole cell extract.
Specifications
Specifications
| Antigen | CITED2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | JA22 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | CITED2 |
| Gene Accession No. | O35740, Q99967 |
| Gene Alias | AI835299; ASD8; Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 2; cbp/p300-interacting transactivator 2; Cbp/p300-interacting transactivator, with Glu/Asp rich carboxy-terminal domain, 2; Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 2; Cited2; ER154-like; melanocyte specific gene 2; melanocyte-specific gene 1-related gene 1; Mrg1; MRG-1; MSG1-related gene 1; Msg2; MSG-related protein 1; P35srj; VSD2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.