Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Citidine Deaminase Protein
SDP

Catalog No. NBP1493065 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1493065 0.5 mg
NBP14930601 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1493065 Supplier R&D Systems™ Supplier No. NBP1493060.5MG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-146 of Human Citidine Deaminase The Recombinant Human Citidine Deaminase Protein is derived from E. coli. The Recombinant Human Citidine Deaminase Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Concentration 0.5mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl Buffer (pH 8.0) containing 1mM DTT, 2mM EDTA, 100mM NaCl, 40% Glycerol
Gene ID (Entrez) 978
Molecular Weight (g/mol) 18.3kDa
Purification Method Protein
Quantity 0.5 mg
Source Human
Immunogen Citidine Deaminase, 1-146aa. MGSSHHHHHHSSGLVPRGSHMAQKRPACTLKPECVQQLLVCSQEAKQSAYCPYSHFPVGAALLTQEGRIFKGCNIENACYPLGICAERTAIQKAVSEGYKDFRAIAIASDMQDDFISPCGACRQVMREFGTNWPVYMTKPDGTYIVMTVQELLPSSFGPEDLQKTQ
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name Citidine Deaminase
Conjugate Unconjugated
Product Line Novus
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.