Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citraconic acid, 99+%

Catalog No. ac110430050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110430050 5 g Glass bottle
AC110430250 25 g Plastic bottle
AC110431000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC110430050 Supplier Thermo Scientific Chemicals Supplier No. 110430050
Only null left
Add to Cart

CAS: 498-23-7 | C5H6O4 | 130.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-23-7
Molecular Formula C5H6O4
Molecular Weight (g/mol) 130.10
MDL Number MFCD00078086
InChI Key HNEGQIOMVPPMNR-IHWYPQMZSA-N
Synonym 2-methylmaleate, citraconate 2-, z-2-methylbut-2-enedioate
PubChem CID 5461090
ChEBI CHEBI:30719
IUPAC Name (Z)-2-methylbut-2-enedioate
SMILES C\C(=C\C(O)=O)C(O)=O

Specifications

Melting Point 88.0°C to 92.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3C(CO2H)=CHCO2H
Quantity 5 g
Beilstein 02,768
Merck Index 15,232
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in alcohol and ether,slightly soluble in chloroform,insoluble in benzene and petroleum ether
Formula Weight 130.1
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Citraconic acid, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-858-7
RTECSNumber GE6650000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.