Learn More
Sigma Organic Chemistry Citraconic anhydride | 25G | 616-02-4 | MFCD00005522 | 0.98
Supplier: Sigma Organic Chemistry 12531825G

Citraconic anhydride, 25G
About This Item:
Synonym(s): 2-Methylmaleic anhydride; 3-Methyl-2,5-furandione; Citraconic acid anhydride; Methylmaleic anhydride; Monomethylmaleic anhydride
Linear Formula: C5H4O3
Molecular Weight: 112.08
CAS Number: 616-02-4
MDL Number: MFCD00005522
UNSPSC Code: 12162002
Purity: 0.98
Boiling Point: 213-214 C (lit.)
Melting Point: 6-10 C (lit.)
Density: 1.247 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.471 (lit.)
Storage: room temp
EINECS Number: 210-459-0
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.