Learn More
Electron Microscopy Sciences Citrajet Low Foaming Liquid Acid Cleaner 1 GL
SDP
Supplier:  Electron Microscopy Sciences 6053201
Encompass_Preferred
Low foaming phosphate-free, concentrated, liquid cleaner and metal brightener, high performance removal of metal oxides, scale, salts and inorganic residues in lab-ware washers, parts washers, power-spray systems and manual and ultrasonic cleaning. Corrosion inhibited and free rinsing leaving no interfering residues. Ideal for acid washing and rinsing. Dilute 2:100 and it has a pH 2.5.
Catalog No. 50-189-8968
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.