Learn More
Electron Microscopy Sciences Citrate Buffer, pH 6 1000 ML
Supplier: Electron Microscopy Sciences 6414208
This buffer is intended for heat-induced antigen retriever on formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies. In IHC most commonly used fixative like formalin mask tissue antigens (cellular, membrane and nuclear) by cross-linking process, this results in poor or no staining in IHC. The use of this buffer on FFPE tissue section improves accessibility of antibodies to tissue antigens.
Intended Use: 1X buffer solution is intended for heat-induced antigen retriever in IHC. Please refer to primary antibody protocol
pH: 6
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.