Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Citrate Synthase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522126
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522126 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522126 Supplier Invitrogen™ Supplier No. PA522126
Only null left
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2, mouse brain, rat brain, Rat muscle. Predicted reactivity: Mouse (94%), Rat (93%), Zebrafish (90%), Xenopus laevis (91%), Pig (95%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a Krebs tricarboxylic acid cycle enzyme that catalyzes the synthesis of citrate from oxaloacetate and acetyl coenzyme A. The enzyme is found in nearly all cells capable of oxidative metabolism. This protein is nuclear encoded and transported into the mitochondrial matrix, where the mature form is found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Citrate Synthase
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene CS
Gene Accession No. O75390, P23007, Q8VHF5, Q9CZU6
Gene Alias 2610511A05Rik; 9030605P22Rik; Ahl4; BB234005; Cis; citrate (Si)-synthase; citrate synthase; citrate synthase precursor (EC 4.1.3.7); citrate synthase, mitochondrial; citrate synthetase; Cs; wu:fb58e04; zgc:55507
Gene Symbols CS
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Citrate synthetase. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100309916, 100309917, 100309924, 100309934, 100310091, 100310549, 100310550, 100310552, 100310659, 100327409, 100327410, 100858903, 12974, 1431, 170587
Target Species Human, Mouse, Rat, Monkey, Chicken
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.