Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric Acid, 1% (w/w), Ricca Chemical
SDP

Catalog No. R2118000500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R2118000500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R2118000500 Supplier Ricca Chemical Company Supplier No. R2118000500A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 77-92-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
MDL Number MFCD00011669
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
PubChem CID 311
ChEBI CHEBI:30769
IUPAC Name water
SMILES O

Specifications

Color Colorless
Quantity 500 mL
Odor Odorless
Packaging Natural Poly Bottle
Physical Form Liquid
Assay Percent Range 1% (w/w)
Percent Purity 1% w/w
Grade Laboratory
Chemical Name or Material Citric Acid
ShelfLife 18 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.