Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric Acid, 99.5 to 100.5% (based on Anhydrous substance), Solstice

Catalog No. 60045754 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
6 x 1 kg
6 x 2.5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-045-754 6 x 2.5 kg
60-022-41 100 g
60-022-42 1 kg
60-045-753 6 x 1 kg
60-022-43 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-045-754 Supplier Solstice Supplier No. 271096X2.5KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Meets analytical specification of BP, Ph. Eur., USP, E330, Anhydrous, 99.5-100.5% (based on anhydrous substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
PubChem CID 311
ChEBI CHEBI:30769
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

Quantity 6 x 2.5 kg
Explosion Limit 8% (65°F)
Residue after Ignition ≤0.05% (as SO4)
Linear Formula HOC(COOH)(CH2COOH)2
UN Number NONH for all modes of transport
Solubility Information Soluble in water, 383g/L at 25°C, Sparingly soluble in Diethyl Ether, Freely soluble in alcohol
Formula Weight 192.12
Percent Purity 99.5 to 100.5% (based on anhydrous substance)
Chemical Name or Material Citric Acid, Meets analytical specification of Ph. Eur.BP, USP
Hazard Statement P305 + P351 + P338
Precautionary Statement H319
EINECSNumber 201-069-1
RTECSNumber GE7350000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.