Learn More
Specifications
Specifications
| Melting Point | 153°C |
| Boiling Point | 175°C |
| Color | White |
| CAS Max % | 100 |
| Density | 1.665g/cm3 |
| Quantity | 500 g |
| Packaging | Poly Bottle |
| Physical Form | Crystalline |
| Specific Gravity | 1.665g/cm3 |
| Linear Formula | C6H8O7 |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves, eye protection. Wash exposed skin thoroughly after handling. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. |
| EINECSNumber | 201-069-1 |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.