Learn More
Citric acid, monosodium salt, 99%, for analysis, anhydrous
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 209.0°C to 215.0°C |
| Assay Percent Range | 99% |
| Packaging | Plastic bottle |
| Linear Formula | HO2CCH2C(OH)(CO2H)CH2CO2Na |
| Quantity | 1 kg |
| Solubility Information | Clear colorless |
| Formula Weight | 214.11 |
| Percent Purity | 99% |
| Grade | Analysis |
| Chemical Name or Material | Citric acid, monosodium salt |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Wash face,hands and any exposed skin thoroughly after handling. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Cont |
| EINECSNumber | 242-734-6 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.