Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric Acid, MP Biomedicals™

Catalog No. 0219464180 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-194-64180 100 g
ICN19464191 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-194-64180 Supplier MP Biomedicals, Inc Supplier No. 0219464180
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Citric Acid acts as an iron chelator.

  • Citric acid can support cholesterol/sterol, ubiquinone and isoprenoid biosynthesis in cell culture applications.
  • In animals, citric acid improves the utilization of nutritional calcium.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

pH 1 to 3 (5% aq. soln.)
Melting Point 149°C to 159°C
Density 1.665 g/cm3 at 18°C
Quantity 100 g
Flash Point 345°C (Closed Cup)
Ignition Point 1010°C (1850°F)
Vapor Pressure Practically zero
Solubility Information Soluble in water (50 g/mL)
Grade Cell Culture Reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.