Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric Acid Stock Solution, 10% w/w Recommended Storage: 2°C to 8°C (36°F to 46°F)., Reagents

Catalog No. 01335597
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-597 250 mL
01-335-598 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-335-597 Supplier Reagents Holdings Llc Supplier No. CC204600250C
Add to Cart
Add to Cart

Citric Acid Stock Solution, 10% w/w, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Citric Acid Stock Solution, 10% w/w
pH <2
CAS 7732-18-5, 77-92-9
Boiling Point 100°C
Color Colorless
Molecular Formula C6H8O7/H2O
Synonym 2-hydroxypropane-1,2,3-tricarboxylic Acid
Percent Purity 10%
Grade Reagent
Quantity 250 mL
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.042
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.