Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric acid, trisodium salt hydrate, 99.8%, for molecular biology, DNAse, RNAse and Protease free

Catalog No. AC327160050
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC327160050 5 kg Plastic bottle
AC327162500 250 g Plastic bottle
AC327160010 1 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC327160050 Supplier Thermo Scientific Chemicals Supplier No. 327160050
Add to Cart

CAS: 114456-61-0 | C6H5Na3O7 | 258.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114456-61-0
Molecular Formula C6H5Na3O7
Molecular Weight (g/mol) 258.07
MDL Number MFCD00150032
InChI Key HRXKRNGNAMMEHJ-UHFFFAOYSA-K
Synonym trisodium citrate hydrate, sodium citrate tribasic hydrate, citric acid trisodium salt hydrate, sodium citrate, tribasic, sodium citrate monohydrate, c6h5o7.3na.h2o, sodium citrate tribasic hydrate, reagentplus r, trisodium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
PubChem CID 21868242
SMILES [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Absorbance 0.02 max. (0.1 M in water) at 260nm
Melting Point >300.0°C
Color White
pH 7 to 9 (5% aq. soln.)
Loss on Drying 12% to 13%
Infrared Spectrum Authentic
Assay Percent Range 0.998
Packaging Plastic bottle
Quantity 5 kg
Merck Index 14, 8602
Solubility Information Solubility in water: soluble.
IUPAC Name trisodium 2-hydroxypropane-1,2,3-tricarboxylate
Formula Weight 258.08
Percent Purity 0.998
Grade Molecular Biology
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Citric acid, trisodium salt hydrate, For molecular biology
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.