Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citrinin, 98%, Thermo Scientific™

Catalog No. AAJ64007LB0
Change view
Click to view available options
Quantity:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64007LB0 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64007LB0 Supplier Thermo Scientific Chemicals Supplier No. J64007LB0

missing translation for 'validMainframeMsg'

Induces mitochondrial permeability

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 518-75-2
Molecular Formula C13H14O5
Quantity 5mg
MDL Number MFCD00006912
UN Number UN3462
Beilstein 5282243
Merck Index 14,2327
Synonym citrinin, antimycin, --citrinin, citriain, 3r,4s-4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acid, unii-3s697x6snz, ccris 175, 3h-2-benzopyran-7-carboxylic acid, 4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-, 3r-trans, 3r-trans-4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acid, 4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acid
InChI Key CBGDIJWINPWWJW-IYSWYEEDSA-N
SMILES CC1C(OC=C2C1=C(C(=C(C2=O)C(=O)O)O)C)C
IUPAC Name (3R,4S)-6-hydroxy-3,4,5-trimethyl-8-oxo-3,4-dihydroisochromene-7-carboxylic acid
Molecular Weight (g/mol) 250.25
PubChem CID 54680783
Formula Weight 250.25
Percent Purity 98%
Chemical Name or Material Citrinin
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H311-H331-H351
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing cancer.
Precautionary Statement GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXINS, EXTRACTED FROM LIVING SOURCES, SOLID, N.O.S.
RTECSNumber DJ2275000
Recommended Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.