Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citrinin, MP Biomedicals™

Catalog No. ICN15070010 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15070010 10 mg
ICN15070001 1 mg
ICN15070005 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15070010 Supplier MP Biomedicals, Inc Supplier No. 0215070010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 518-75-2
Molecular Formula C13H14O5
Molecular Weight (g/mol) 250.25
InChI Key CBGDIJWINPWWJW-IYSWYEEDSA-N
Synonym citrinin, antimycin, --citrinin, citriain, 3r,4s-4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acid, unii-3s697x6snz, ccris 175, 3h-2-benzopyran-7-carboxylic acid, 4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-, 3r-trans, 3r-trans-4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acid, 4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acid
PubChem CID 54680783
IUPAC Name (3R,4S)-6-hydroxy-3,4,5-trimethyl-8-oxo-3,4-dihydroisochromene-7-carboxylic acid
SMILES CC1C(OC=C2C1=C(C(=C(C2=O)C(=O)O)O)C)C

Specifications

Color Yellow
Quantity 10 mg
Formula Weight 250.3
Physical Form Crystalline Powder
Chemical Name or Material Citrinin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.