Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATCC Citrobacter freundii (ATCC BAA-3249) Frozen Drug-Resistant Strain 990954, BSL-2

Supplier:  ATCC BAA3249

Encompass

Citrobacter freundii strain 990954 is a drug-resistant clinical isolate recovered from a human skin ulcer in Brazil (2013). Supplied as a frozen culture for research use, this strain supports antimicrobial resistance studies, drug development, molecular assay validation, and comparative genomics.

  • Product format: Frozen.
  • Strain designation: 990954.
  • Genome sequenced strain: Yes.
  • Isolation source: Skin ulcer (human).
  • Geographical isolation: Brazil.
  • Applications: Antimicrobial resistance research, bioinformatics, drug development, assay validation.
  • Storage conditions: -80°C or colder.
  • Biosafety level: BSL 2.
  • Suggested medium: Trypticase soy agar/broth.
  • Incubation temperature: 37°C.
  • Atmosphere: Aerobic.
  • Gram stain: Negative.
  • Packaging size: Not specified on manufacturer page.
  • Concentration: Not specified on manufacturer page.

Catalog No. NC3538698


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.