Learn More
ATCC Citrobacter freundii (ATCC BAA-3295) - Drug-resistant clinical isolate, frozen culture
Supplier:  ATCC BAA3295
Encompass
Citrobacter freundii strain BAA-3295 is a multidrug-resistant clinical isolate recovered from bronchoalveolar lavage in 2013. Supplied as a frozen culture, it is intended for research into antimicrobial resistance, molecular detection assay development, and sterility testing. The strain exhibits resistance to multiple antibiotics while remaining susceptible to selected agents per the manufacturer's antibiogram.
  • Isolated from respiratory bronchoalveolar lavage (2013).
  • Supplied as a frozen culture; store at -80°C or colder.
  • Grows on trypticase soy agar/broth at 37°C under aerobic conditions.
  • Displays a multidrug-resistant antimicrobial profile with susceptibility to selected agents.
  • Intended for research use in antimicrobial development and assay validation.
Catalog No. NC3538874