Learn More
ATCC Citrobacter freundii (Braak) Werkman and Gillen - ATCC 8454, freeze-dried reference strain
Supplier:  ATCC 8454/DEL
Encompass
Citrobacter freundii (Braak) Werkman and Gillen is a reference bacterial strain supplied for laboratory research and quality control. The strain is provided as a freeze-dried culture with documented growth conditions and recommended maintenance media, and it is used in studies involving glycerol degradation and trimethylene glycol production.
  • Available as a freeze-dried culture for controlled rehydration and propagation.
  • Stable when stored at 2°C to 8°C to maintain viability.
  • Grows aerobically at 37°C under standard laboratory conditions.
  • Propagate on nutrient agar or nutrient broth (ATCC Medium 3) for routine culture.
  • Useful for research into glycerol degradation and production of trimethylene glycol.
  • Includes documented strain aliases and provenance for traceability in testing.
Catalog No. NC3876621